Assay Details
| Gene Symbol: KCNQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004700.3 | NP_004691.2 | 9 - 10 | 1377 | 62 | |
| XM_017002792.1 | XP_016858281.1 | 6 - 7 | 618 | 62 | ||
| GenBank mRNA | AF105202.1 | - | 9 - 10 | 1377 | 62 | |
| AK074957.1 | - | 6 - 7 | 614 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000518.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
sensory perception of sound
negative regulation of synaptic transmission, dopaminergic
regulation of ion transmembrane transport
inner ear morphogenesis
potassium ion transmembrane transport
delayed rectifier potassium channel activity
potassium channel activity
protein binding
potassium ion transport
sensory perception of sound
negative regulation of synaptic transmission, dopaminergic
regulation of ion transmembrane transport
inner ear morphogenesis
potassium ion transmembrane transport
delayed rectifier potassium channel activity
potassium channel activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | ENST00000347132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | ENST00000509682.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01008134.hg.1 | TC01000518.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100156201.hg.1 | TC0100007971.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000347132.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000443478.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000506017.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | ENST00000509682.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_004700.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007971.hg.1 | NM_172163.2 | Refseq |

KCNQ4
Genomic Map