| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003898.hg.1 |
ENST00000062104.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003898.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20003898.hg.1 |
TC20000274.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150354.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150354.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150354.hg.1 |
TC2000007288.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000150354.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150354.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000062104.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
ENST00000346199.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_005386.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007288.hg.1 |
NM_181689.2 |
Refseq |