| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21005877.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21005877.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21005877.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR21005877.hg.1 |
TC21000485.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008250.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
NM_005423.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
TC2100008250.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000291526.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000463771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
ENST00000475297.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100155897.hg.1 |
NM_005423.4 |
Refseq |