| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000289893.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000317713.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000361689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000372900.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000372915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000476350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000539005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
ENST00000545844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
NM_012090.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007607.hg.1 |
TC01000493.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000289893.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000317713.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000361689.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000372900.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000372915.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000476350.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000539005.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
ENST00000545844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
NM_012090.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007606.hg.1 |
TC01000493.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000289893.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000361689.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000372915.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000372925.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000476350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000530262.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000564288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000567887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
NM_012090.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000289893.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000361689.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000372915.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000372925.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000476350.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000530262.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000564288.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
ENST00000567887.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
NM_012090.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100155548.hg.1 |
TC0100007898.hg.1 |
Clariom D TC |