| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001138.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001138.hg.1 |
ENST00000427562.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001138.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001138.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20001138.hg.1 |
TC20000077.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
TC2000006676.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000145805.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000246070.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
ENST00000427562.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_001199897.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006676.hg.1 |
NM_012261.3 |
Refseq |