| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003542.hg.1 |
ENST00000221576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003542.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003542.hg.1 |
TC19000240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008914.hg.1 |
ENST00000424331.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003543.hg.1 |
ENST00000221576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003543.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003543.hg.1 |
TC19000240.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003541.hg.1 |
ENST00000221576.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003541.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19003541.hg.1 |
TC19000240.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
ENST00000593274.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011675.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199558.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199558.hg.1 |
TC1900011675.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199558.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
TC1900011675.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000588841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
ENST00000592760.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199566.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199561.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199561.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199561.hg.1 |
TC1900011675.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199561.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199561.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008914.hg.1 |
ENST00000424331.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
NM_014047.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
TC1900011675.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
ENST00000221576.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
ENST00000588858.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900199562.hg.1 |
NM_014047.2 |
Refseq |