| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020596.hg.1 |
ENST00000306185.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020596.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08020596.hg.1 |
TC08001564.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518696.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000520677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000521648.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000522717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
ENST00000613356.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800011633.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
ENST00000306185.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
ENST00000518918.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
NM_014078.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800179002.hg.1 |
TC0800011633.hg.1 |
Clariom D TC |