| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000262829.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000338644.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000359369.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000397303.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000405409.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000414461.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000416721.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000438146.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
ENST00000449924.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_001031695.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_001082576.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_001082577.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_001082578.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_001082579.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
NM_014309.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012271.hg.1 |
TC22000699.hg.1 |
HTA TC |