Assay Details
| Gene Symbol: TNFAIP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF099936.1 | - | 1 - 2 | 92 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF099936.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000274456.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000388882.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000415806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000503646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504642.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000504771.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | ENST00000513374.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001077654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286813.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286815.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_001286817.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008437.hg.1 | NM_014350.3 | Refseq |

TNFAIP8
Genomic Map