| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021722.hg.1 |
ENST00000244333.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021722.hg.1 |
ENST00000377995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021722.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021722.hg.1 |
TC19001586.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021723.hg.1 |
ENST00000244333.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021723.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021723.hg.1 |
TC19001586.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021721.hg.1 |
ENST00000244333.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021721.hg.1 |
ENST00000377995.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021721.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19021721.hg.1 |
TC19001586.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
TC1900010847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189911.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189916.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189916.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189916.hg.1 |
TC1900010847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189916.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189916.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
TC1900010847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900189912.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000244333.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000594326.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000595970.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
ENST00000597741.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010847.hg.1 |
NM_014400.2 |
Refseq |