| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
ENST00000394210.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
ENST00000412050.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10019158.hg.1 |
TC10001526.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000177522.hg.1 |
TC1000011420.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000265997.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000412050.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
ENST00000614585.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_001178137.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011420.hg.1 |
NM_014912.4 |
Refseq |