Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016071.3 | NP_057155.1 | 1 - 2 | 134 | 93 | |
| GenBank mRNA | AF151897.1 | - | 1 - 2 | 112 | 93 | |
| AK024776.1 | - | 1 - 2 | 132 | 93 | ||
| BP870829.1 | - | 1 - 2 | 133 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001928.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF151897.1,NM_016071.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | ENST00000610021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | ENST00000324787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | ENST00000393008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | ENST00000496958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | ENST00000544013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029975.hg.1 | TC07001928.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029980.hg.1 | ENST00000393008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029980.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029980.hg.1 | TC07001928.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400010426.hg.1 | ENST00000507850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029978.hg.1 | ENST00000393008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029978.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029978.hg.1 | TC07001928.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000324787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000393008.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000496958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | ENST00000544013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029974.hg.1 | TC07001928.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0700192498.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192498.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192498.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192498.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192498.hg.1 | TC0700012824.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192496.hg.1 | TC0700012824.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400010426.hg.1 | ENST00000507850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000472343.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000484502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | ENST00000610021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700012824.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192493.hg.1 | TC0700012824.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000324787.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000393008.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000467334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000469351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000485202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000496641.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | ENST00000496958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | NM_016071.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | NM_053035.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700192491.hg.1 | TC0700012824.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 173286 | - | - |

MRPS33
Genomic Map