| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014743.hg.1 |
ENST00000338272.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014743.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014743.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014743.hg.1 |
TC01000937.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014745.hg.1 |
ENST00000338272.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014745.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014745.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01014745.hg.1 |
TC01000937.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100009339.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000338272.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000473828.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
ENST00000479160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
NM_020141.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100166554.hg.1 |
TC0100009339.hg.1 |
Clariom D TC |