Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024667.2 | NP_078943.1 | 1 - 2 | 215 | 74 | |
| GenBank mRNA | AK022812.1 | - | 1 - 2 | 215 | 74 | |
| BC005882.2 | - | 1 - 2 | 153 | 3533384 | 74 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002080.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
autophagy
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
intracellular transport of virus
positive regulation of viral release from host cell
positive regulation of viral budding via host ESCRT complex
protein binding
calcium-dependent protein binding
autophagy
protein transport
endosomal transport
viral life cycle
multivesicular body assembly
viral budding via host ESCRT complex
intracellular transport of virus
positive regulation of viral release from host cell
positive regulation of viral budding via host ESCRT complex
protein binding
calcium-dependent protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12027394.hg.1 | ENST00000267202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027394.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027394.hg.1 | TC12002080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027395.hg.1 | ENST00000267202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027395.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027395.hg.1 | TC12002080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027396.hg.1 | ENST00000267202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027396.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12027396.hg.1 | TC12002080.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012254.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000371248.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000543590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | ENST00000623210.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195914.hg.1 | TC1200012254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195924.hg.1 | TC1200012254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | ENST00000535765.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195923.hg.1 | TC1200012254.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200195926.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195926.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195926.hg.1 | ENST00000267202.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200195926.hg.1 | NM_024667.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200195926.hg.1 | TC1200012254.hg.1 | Clariom D TC |

VPS37B
Genomic Map