Assay Details
| Gene Symbol: NYNRIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021873.1 | - | 2 - 3 | 133 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK021873.1 transcripts.
Set Membership: |
|

NYNRIN
Genomic Map