| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000240050.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000392830.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000547115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000547115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000547115.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000548101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000548101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000548101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000548101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000548101.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550496.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000550736.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
ENST00000552029.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_001033050.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011811.hg.1 |
NM_025198.4 |
Refseq |