Assay Details
| Gene Symbol: BMP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001719.2 | NP_001710.1 | 2 - 3 | 1139 | 73 | |
| GenBank mRNA | AK291186.1 | - | 2 - 3 | 715 | 73 | |
| AK312419.1 | - | 2 - 3 | 712 | 73 | ||
| BC008584.1 | - | 2 - 3 | 651 | 4183402 | 73 | |
| EU679366.1 | - | 2 - 3 | 523 | 73 | ||
| M60316.1 | - | 2 - 3 | 706 | 73 | ||
| X51801.1 | - | 2 - 3 | 732 | 73 | ||
More Information
Set Membership: |
|

BMP7
Genomic Map