Assay Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022347.3 | NP_071742.1 | 3 | 1128 | 133 | |
| XM_017000477.1 | XP_016855966.1 | 2 | 1801 | 133 | ||
| XM_017000478.1 | XP_016855967.1 | 2 | 1134 | 133 | ||
| GenBank mRNA | AJ299406.1 | - | 1 | 201 | 133 | |
| AK289793.1 | - | 3 | 1103 | 133 | ||
| BC096704.1 | - | 1 | 201 | 40021613 | 133 | |
| BC098170.1 | - | 1 | 201 | 40021614 | 133 | |
| BC098309.1 | - | 1 | 201 | 40021617 | 133 | |
| BC098348.1 | - | 1 | 201 | 40021612 | 133 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003588.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055319.hg.1 | ENST00000482587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055319.hg.1 | ENST00000553856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055319.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055319.hg.1 | TC01003588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055320.hg.1 | ENST00000482587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055320.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055320.hg.1 | ENST00000553856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055320.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055320.hg.1 | TC01003588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055321.hg.1 | ENST00000482587.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055321.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055321.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055321.hg.1 | TC01003588.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225829.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225829.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225829.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225829.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225829.hg.1 | TC0100016567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225833.hg.1 | TC0100016567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000367612.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000474318.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000483123.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | ENST00000609928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_001199260.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016567.hg.1 | NM_145034.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225830.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225830.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225830.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225830.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225830.hg.1 | TC0100016567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225831.hg.1 | TC0100016567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | ENST00000482587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | ENST00000495650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | NM_022347.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100225832.hg.1 | TC0100016567.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 113764 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 711908 | - | - |

TOR1AIP2
Genomic Map