| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8008701 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8008700 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
220529_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700008379.hg.1 |
ENST00000623762.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700008379.hg.1 |
ENST00000623762.1 |
Ensembl |
| Human Genome U133 Plus 2.0 Array |
220529_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
220529_at |
- |
- |
| Human Genome U133A 2.0 Array |
220529_at |
- |
- |
| Human Genome U133A 2.0 Array |
220529_PM_at |
- |
- |
| AffymetrixPathwork Diagnostics Pathchip |
pw534345 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at4 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
220529_at6 |
- |
- |
| Human Genome U95 Set |
67414_f_at |
- |
- |
| Human Genome U95 Set |
67414_PM_f_at |
- |
- |
| Human Genome U133 Set |
220529_at |
- |
- |
| Human Genome U133 Set |
220529_PM_at |
- |
- |