| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020559.hg.1 |
TC02001322.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020559.hg.1 |
ENST00000490341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020559.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020561.hg.1 |
TC02001322.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020561.hg.1 |
ENST00000490341.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02020561.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200177494.hg.1 |
TC0200010839.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200177496.hg.1 |
TC0200010839.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000473885.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000485041.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
ENST00000490341.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200010839.hg.1 |
NR_003063.1 |
Refseq |