Assay Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032051.1 | NP_114440.1 | 2 - 3 | 1996 | 98 | |
| GenBank mRNA | AF119256.1 | - | 2 - 3 | 1995 | 98 | |
| AF254082.1 | - | 2 - 3 | 1965 | 98 | ||
| AK123441.2 | - | 1 - 2 | 695 | 98 | ||
| AK291803.1 | - | 2 - 3 | 1509 | 98 | ||
| BC021091.1 | - | 2 - 3 | 1543 | 4857537 | 98 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000668.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
male gonad development
T cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
metal ion binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
spermatogenesis
male gonad development
T cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011799.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011799.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011799.hg.1 | TC22000668.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000266269.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000351933.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000405309.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | ENST00000494109.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_014323.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032050.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008477.hg.1 | NM_032052.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | TC2200008477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160986.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | NM_032051.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | TC2200008477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | ENST00000215919.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | ENST00000465287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160987.hg.1 | NM_032051.1 | Refseq |

PATZ1
Genomic Map