Assay Details
| Gene Symbol: MAGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032121.5 | NP_115497.4 | 1 - 2 | 260 | 67 | |
| GenBank mRNA | AK223131.1 | - | 1 - 2 | 123 | 67 | |
| AK294943.1 | - | 1 - 2 | 285 | 67 | ||
| AK296743.1 | - | 1 - 2 | 123 | 67 | ||
| AK314309.1 | - | 1 - 2 | 123 | 67 | ||
| AL136636.1 | - | 1 - 2 | 67 | |||
| AY358691.1 | - | 1 - 2 | 110 | 67 | ||
| BC041014.1 | - | 1 - 2 | 107 | 3834858 | 67 | |
| BC060842.1 | - | 1 - 2 | 123 | 30338916 | 67 | |
| BC063037.1 | - | 1 - 2 | 132 | 67 | ||
| DC329979.1 | - | 1 - 2 | 260 | 67 | ||
| DQ000004.1 | - | 1 - 2 | 67 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001174.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015825.hg.1 | ENST00000358075.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015825.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015825.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015825.hg.1 | TC0X001174.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015826.hg.1 | ENST00000358075.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015826.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015826.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015826.hg.1 | TC0X001174.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015830.hg.1 | ENST00000358075.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015830.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015830.hg.1 | TC0X001174.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169180.hg.1 | TC0X00010130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169185.hg.1 | TC0X00010130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169179.hg.1 | TC0X00010130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00169186.hg.1 | TC0X00010130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000358075.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000373336.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000476168.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000610432.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | ENST00000618282.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010130.hg.1 | NM_032121.5 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 187717 | - | - |

MAGT1
Genomic Map