| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
ENST00000258499.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
ENST00000393091.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12024145.hg.1 |
TC12001835.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000258499.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000393091.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
ENST00000537435.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_001042403.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_001278393.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NM_032147.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
NR_125360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189775.hg.1 |
TC1200011588.hg.1 |
Clariom D TC |