| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010071.hg.1 |
ENST00000335288.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010071.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010071.hg.1 |
TC13000897.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010072.hg.1 |
ENST00000335288.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010072.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13010072.hg.1 |
TC13000897.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300009919.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166046.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166046.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166046.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166046.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166046.hg.1 |
TC1300009919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300166044.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166044.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166044.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166044.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166044.hg.1 |
TC1300009919.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300166045.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166045.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166045.hg.1 |
ENST00000335288.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300166045.hg.1 |
NM_000705.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300166045.hg.1 |
TC1300009919.hg.1 |
Clariom D TC |