| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005700.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005700.hg.1 |
TC20000399.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005703.hg.1 |
ENST00000303004.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005703.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005703.hg.1 |
TC20000399.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005701.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20005701.hg.1 |
TC20000399.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
ENST00000613921.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009418.hg.1 |
NR_125739.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
TC2000007682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153566.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007682.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
TC2000007682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153569.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
TC2000007682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153570.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
TC2000007682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153571.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_005194.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
TC2000007682.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
ENST00000303004.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_001285878.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_001285879.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000153567.hg.1 |
NM_005194.3 |
Refseq |