| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002837.hg.1 |
ENST00000268719.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002837.hg.1 |
ENST00000376345.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002837.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002837.hg.1 |
TC17000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000579871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
ENST00000579871.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002838.hg.1 |
ENST00000268719.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002838.hg.1 |
ENST00000376345.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002838.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002838.hg.1 |
TC17000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002836.hg.1 |
ENST00000268719.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002836.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002836.hg.1 |
TC17000203.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002835.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002835.hg.1 |
TC17000203.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
TC1700007097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149989.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149987.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149987.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149987.hg.1 |
TC1700007097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700149987.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149987.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
TC1700007097.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700149990.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000268719.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000376345.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000459843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000481836.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000579871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
ENST00000579871.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007097.hg.1 |
NM_024052.4 |
Refseq |