| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Gene 1.0 ST Array |
8070631 |
- |
- |
| Affymetrix Human Gene 1.0 ST Array |
8070630 |
- |
- |
| Affymetrix Human Gene 1.1 ST Array |
8070629 |
- |
- |
| Affymetrix HT HG-U133+ PM Array Plate |
232191_PM_at |
- |
- |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012273.hg.1 |
ENST00000417786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012273.hg.1 |
ENST00000417786.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100008279.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007709.hg.1 |
ENST00000624242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012273.hg.1 |
ENST00000417786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012273.hg.1 |
ENST00000417786.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100008279.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
TC2100008279.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156096.hg.1 |
ENST00000617971.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007709.hg.1 |
ENST00000624242.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156098.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156098.hg.1 |
NM_001308491.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2100156098.hg.1 |
TC2100008279.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2100156098.hg.1 |
ENST00000447535.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2100156098.hg.1 |
NM_001308491.1 |
Refseq |
| Human Genome U133 Plus 2.0 Array |
232191_PM_at |
- |
- |
| Human Genome U133 Plus 2.0 Array |
232191_at |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at2 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at10 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at1 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at3 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at5 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at8 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at11 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at9 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at6 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at7 |
- |
- |
| Affymetrix Human Genome U133 Plus 2.0 Array |
232191_at4 |
- |
- |
| Human Genome U95 Set |
65024_PM_at |
- |
- |
| Human Genome U95 Set |
65024_at |
- |
- |
| Human Genome U133 Set |
232191_at |
- |
- |
| Human Genome U133 Set |
240920_PM_at |
- |
- |
| Human Genome U133 Set |
232191_PM_at |
- |
- |
| Human Genome U133 Set |
240920_at |
- |
- |