Assay Details
| Gene Symbol: ZBTB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164342.2 | NP_001157814.1 | 5 | 19344 | 81 | |
| NM_001164343.2 | NP_001157815.1 | 12 | 19806 | 81 | ||
| NM_001164344.2 | NP_001157816.1 | 8 | 19770 | 81 | ||
| NM_001164345.2 | NP_001157817.1 | 7 | 19724 | 81 | ||
| NM_001164346.2 | NP_001157818.1 | 6 | 19363 | 81 | ||
| NM_001164347.2 | NP_001157819.1 | 6 | 19385 | 81 | ||
| NM_015642.5 | NP_056457.3 | 10 | 19568 | 81 | ||
| NR_121662.1 | 7 | 17826 | 81 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZBTB20
Genomic Map