Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003179.2 | NP_003170.1 | 3 - 4 | 244 | 63 | |
| GenBank mRNA | AK313030.1 | - | 3 - 4 | 229 | 63 | |
| BC064550.1 | - | 3 - 4 | 244 | 5768012 | 63 | |
| X06389.1 | - | 2 - 3 | 177 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001029.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
synaptic vesicle maturation
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
regulation of short-term neuronal synaptic plasticity
synaptic vesicle membrane organization
cellular response to organic substance
regulation of opioid receptor signaling pathway
transporter activity
cholesterol binding
SH2 domain binding
identical protein binding
protein self-association
endocytosis
synaptic vesicle maturation
regulation of neuronal synaptic plasticity
regulation of long-term neuronal synaptic plasticity
regulation of short-term neuronal synaptic plasticity
synaptic vesicle membrane organization
cellular response to organic substance
regulation of opioid receptor signaling pathway
transporter activity
cholesterol binding
SH2 domain binding
identical protein binding
protein self-association
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013812.hg.1 | ENST00000263233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013812.hg.1 | ENST00000479808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013812.hg.1 | ENST00000538567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013812.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013812.hg.1 | TC0X001029.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013813.hg.1 | ENST00000263233.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013813.hg.1 | ENST00000479808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013813.hg.1 | ENST00000538567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013813.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X013813.hg.1 | TC0X001029.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165974.hg.1 | TC0X00009661.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472598.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000472737.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | ENST00000494396.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009661.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000263233.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000376303.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000466635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000469389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | ENST00000479808.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | NM_003179.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00165975.hg.1 | TC0X00009661.hg.1 | Clariom D TC |
Citations (1)
Altered neuronal gene expression in brain regions differentially affected by Alzheimer's disease: a reference data set.
- Authors
- Liang WS, Dunckley T, Beach TG, Grover A, Mastroeni D, Ramsey K, Caselli RJ, Kukull WA, McKeel D, Morris JC, Hulette CM, Schmechel D, Reiman EM, Rogers J, Stephan DA
- Journal
- Physiol Genomics - Page No 240 (Tue Apr 22 00:00:00 UTC 2008)
- Volume (Year)
- 33 (2008)
- Product Usage
- -

SYP
Genomic Map