Assay Details
| Gene Symbol: HCST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017026193.1 | XP_016881682.1 | 3 | 107 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000246551.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | ENST00000437550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_001007469.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007891.hg.1 | NM_014266.3 | Refseq |

HCST
Genomic Map