Assay Details
| Gene Symbol: LOC146795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135222.1 | 1 - 2 | 1330 | 55 | ||
| GenBank mRNA | AK057377.1 | - | 1 - 2 | 1202 | 55 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451804 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035315 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013615 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451936 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451821 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035220 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035309 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451841 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451941 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451946 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035225 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035169 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451921 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451767 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035234 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451873 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451779 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451909 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035142 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451944 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035195 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451939 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035272 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035260 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035168 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451927 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451872 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451819 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451828 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035277 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451910 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451800 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451906 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035258 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451907 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013621 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451801 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451838 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451919 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013625 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451857 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451902 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035240 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451898 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451924 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451891 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451846 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451862 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451879 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451861 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451950 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035329 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035219 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451774 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451914 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451874 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451794 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013624 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035311 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451896 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451952 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451777 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451863 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451884 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451775 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451797 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451887 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013626 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451840 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451881 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035317 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035189 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035265 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451822 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013632 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035259 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451851 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451945 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035182 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035268 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451776 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451829 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451953 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451813 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451766 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451860 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451916 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013645 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451923 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17013627 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451842 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451866 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451782 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035143 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035237 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035303 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10451948 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31035247 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 271908 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | AB209879.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | AK125700.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | AL832238.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | BC016834.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | BC037169.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | NM_000935.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | NM_182943.2 | refseq_mrna_human_NM |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 176082 | U84573.1 | genbank_mrna_human |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 53592 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 870729 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3494109 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4061 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3494109 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 123138 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03286738 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03286733 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03286726 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03286733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1564402_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008797.hg.1 | ENST00000577573.1 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 1564402_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564402_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564402_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 234525 | - | - |

LOC146795
Genomic Map