| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500006711.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500145941.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500145941.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500145941.hg.1 |
ENST00000510879.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500145941.hg.1 |
NR_109935.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500145941.hg.1 |
TC0500006711.hg.1 |
Clariom D TC |