Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005268380.2 | XP_005268437.1 | 8 - 9 | 1809 | 95 | |
| XM_011537567.2 | XP_011535869.1 | 8 - 9 | 1809 | 95 | ||
| XM_011537568.2 | XP_011535870.1 | 8 - 9 | 1803 | 95 | ||
| GenBank mRNA | AK058046.1 | - | 2 - 3 | 295 | 95 | |
| BC031650.2 | - | 1 - 2 | 253 | 95 | ||
| BC073914.1 | - | 2 - 3 | 281 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK058046.1, BC073914.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000359120.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000503848.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000504522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000506591.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000509286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | ENST00000511705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500008994.hg.1 | NM_152550.3 | Refseq |

SH3RF2
Genomic Map