Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021979.3 | NP_068814.2 | 1 | 1554 | 74 | |
| GenBank mRNA | AK097824.1 | - | 2 | 603 | 74 | |
| BC001752.1 | - | 2 | 1248 | 3358246 | 74 | |
| BC036107.1 | - | 1 | 1280 | 5276121 | 74 | |
| BT009815.1 | - | 1 | 1172 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

HSPA2
Genomic Map