| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012678.hg.1 |
ENST00000249042.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012678.hg.1 |
ENST00000403892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012678.hg.1 |
ENST00000413912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012678.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012678.hg.1 |
TC22000718.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012674.hg.1 |
ENST00000249042.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012674.hg.1 |
ENST00000403892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012674.hg.1 |
ENST00000413912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012674.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012674.hg.1 |
TC22000718.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012675.hg.1 |
ENST00000249042.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012675.hg.1 |
ENST00000403892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012675.hg.1 |
ENST00000413912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012675.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012675.hg.1 |
TC22000718.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012676.hg.1 |
ENST00000249042.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012676.hg.1 |
ENST00000403892.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012676.hg.1 |
ENST00000413912.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012676.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012676.hg.1 |
TC22000718.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008630.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
TC2200008630.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162255.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
TC2200008630.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162260.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
TC2200008630.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162256.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
NM_003312.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
TC2200008630.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000249042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000403892.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000438203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
ENST00000622841.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
NM_001270483.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200162257.hg.1 |
NM_003312.5 |
Refseq |