Assay Details
| Gene Symbol: SLC7A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032803.5 | NP_116192.4 | 1 - 2 | 147 | 66 | |
| GenBank mRNA | AF320612.1 | - | 1 - 2 | 98 | 66 | |
| AK074865.1 | - | 1 - 2 | 118 | 66 | ||
| BC033816.1 | - | 1 - 2 | 102 | 5206252 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001117.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF320612.1,AK074865.1,BC033816.1,NM_032803.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transport
arginine transport
lysine transport
ornithine transport
regulation of TOR signaling
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
basic amino acid transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity
amino acid transport
arginine transport
lysine transport
ornithine transport
regulation of TOR signaling
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
basic amino acid transmembrane transport
L-ornithine transmembrane transporter activity
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015213.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015213.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015213.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015213.hg.1 | TC0X001117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015211.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015211.hg.1 | ENST00000374299.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015211.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015211.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X015211.hg.1 | TC0X001117.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168101.hg.1 | TC0X00009972.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | ENST00000374299.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00009972.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | ENST00000298085.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | NM_001048164.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | NM_032803.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00168103.hg.1 | TC0X00009972.hg.1 | Clariom D TC |

SLC7A3
Genomic Map