| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000359918.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397067.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000397068.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000425767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000425767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000425767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000425767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
ENST00000453353.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001079536.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_001297668.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NM_053017.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100009855.hg.1 |
NR_123732.1 |
Refseq |