Assay Details
| Gene Symbol: IL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014439.3 | NP_055254.2 | 2 - 3 | 191 | 134 | |
| XM_011510963.2 | XP_011509265.1 | 3 - 4 | 2192 | 134 | ||
| GenBank mRNA | AF167368.1 | - | 2 - 3 | 191 | 134 | |
| AF200496.1 | - | 2 - 3 | 197 | 134 | ||
| AF251118.1 | - | 2 - 3 | 221 | 134 | ||
| AF251119.1 | - | 2 - 3 | 149 | 134 | ||
| BC020637.1 | - | 2 - 3 | 152 | 4731213 | 134 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000714.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | ENST00000263326.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | ENST00000352179.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02011088.hg.1 | TC02000714.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000349806.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | ENST00000353225.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173203.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173204.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008942.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000263326.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000311328.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | ENST00000352179.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_014439.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_173202.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | NM_173205.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200162447.hg.1 | TC0200008942.hg.1 | Clariom D TC |

IL37
Genomic Map