| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000327028.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000343765.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000448935.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000553888.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000556412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
ENST00000557658.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
NM_001193314.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
NM_001193315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
NM_001193316.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
NM_001193317.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
NM_022067.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14015283.hg.1 |
TC14001346.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000327028.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000343765.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000448935.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553576.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553888.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000556412.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000557658.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_022067.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000327028.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000343765.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000448935.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553576.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553691.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000553888.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000556412.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
ENST00000557658.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193314.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193316.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_001193317.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
NM_022067.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400172204.hg.1 |
TC1400009784.hg.1 |
Clariom D TC |