Assay Details
| Gene Symbol: HHIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022475.2 | NP_071920.1 | 10 - 11 | 2355 | 87 | |
| XM_005263178.4 | XP_005263235.1 | 10 - 11 | 2224 | 87 | ||
| XM_006714288.3 | XP_006714351.1 | 10 - 11 | 2224 | 87 | ||
| GenBank mRNA | AK024645.1 | - | 3 - 4 | 342 | 87 | |
| AK291849.1 | - | 10 - 11 | 1695 | 87 | ||
| AY009317.1 | - | 10 - 11 | 2195 | 87 | ||
| AY009951.1 | - | 10 - 11 | 2330 | 87 | ||
| AY358747.1 | - | 10 - 11 | 2305 | 87 | ||
| BC025311.1 | - | 10 - 11 | 2181 | 4122618 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000706.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK024645.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
carbohydrate metabolic process
smoothened signaling pathway
neuroblast proliferation
dorsal/ventral pattern formation
negative regulation of signal transduction
regulation of fibroblast growth factor receptor signaling pathway
negative regulation of smoothened signaling pathway
skeletal system morphogenesis
oxidation-reduction process
epithelial tube branching involved in lung morphogenesis
protein binding
zinc ion binding
oxidoreductase activity, acting on the CH-OH group of donors, quinone or similar compound as acceptor
quinone binding
hedgehog family protein binding
carbohydrate metabolic process
smoothened signaling pathway
neuroblast proliferation
dorsal/ventral pattern formation
negative regulation of signal transduction
regulation of fibroblast growth factor receptor signaling pathway
negative regulation of smoothened signaling pathway
skeletal system morphogenesis
oxidation-reduction process
epithelial tube branching involved in lung morphogenesis
protein binding
zinc ion binding
oxidoreductase activity, acting on the CH-OH group of donors, quinone or similar compound as acceptor
quinone binding
hedgehog family protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04010044.hg.1 | ENST00000296575.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010044.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04010044.hg.1 | TC04000706.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000503090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000503090.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000434550.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000503090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000503090.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000505891.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000509630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000511314.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000512791.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | ENST00000515080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008892.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161076.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161076.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161076.hg.1 | ENST00000296575.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400161076.hg.1 | NM_022475.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400161076.hg.1 | TC0400008892.hg.1 | Clariom D TC |

HHIP
Genomic Map