| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014969.hg.1 |
ENST00000216264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014969.hg.1 |
ENST00000541677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014969.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014969.hg.1 |
TC22000890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014968.hg.1 |
ENST00000216264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014968.hg.1 |
ENST00000541677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014968.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014968.hg.1 |
TC22000890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014970.hg.1 |
ENST00000216264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014970.hg.1 |
ENST00000541677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014970.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014970.hg.1 |
TC22000890.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000460254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000460254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014966.hg.1 |
ENST00000216264.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014966.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22014966.hg.1 |
TC22000890.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000460254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000460254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
ENST00000471929.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200009028.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
TC2200009028.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165406.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165405.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165405.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165405.hg.1 |
TC2200009028.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165405.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165405.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
TC2200009028.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165409.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
TC2200009028.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165407.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
NM_022766.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
TC2200009028.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
ENST00000216264.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
ENST00000443629.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200165408.hg.1 |
NM_022766.5 |
Refseq |