Assay Details
| Gene Symbol: CLYBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206808.3 | NP_996531.1 | 4 - 5 | 572 | 68 | |
| NR_104592.1 | 4 - 5 | 572 | 68 | |||
| XM_005254030.2 | XP_005254087.1 | 4 - 5 | 551 | 68 | ||
| XM_006719915.3 | XP_006719978.1 | 4 - 5 | 551 | 68 | ||
| XM_011521051.2 | XP_011519353.1 | 4 - 5 | 551 | 68 | ||
| XM_011521052.2 | XP_011519354.1 | 4 - 5 | 551 | 68 | ||
| GenBank mRNA | AF428253.1 | - | 4 - 5 | 540 | 68 | |
| AK095506.1 | - | 4 - 5 | 549 | 68 | ||
| BC034360.1 | - | 4 - 5 | 554 | 4750050 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000358.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | ENST00000339105.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | ENST00000376354.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | ENST00000376355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | ENST00000376360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | ENST00000444838.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003784.hg.1 | TC13000358.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152814.hg.1 | TC1300007845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |

CLYBL
Genomic Map