Assay Details
| Gene Symbol: SPATA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139073.3 | NP_620712.2 | 2 - 3 | 579 | 90 | |
| XM_005246303.2 | XP_005246360.1 | 2 - 3 | 577 | 90 | ||
| XM_017003363.1 | XP_016858852.1 | 2 - 3 | 577 | 90 | ||
| GenBank mRNA | AK093385.1 | - | 2 - 3 | 577 | 90 | |
| AY032925.1 | - | 3 - 4 | 677 | 90 | ||
| BC047704.1 | - | 3 - 4 | 522 | 5744971 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001384.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000355662.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000424440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000433428.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000452881.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02021708.hg.1 | TC02001384.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000495639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011048.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000409956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000423134.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000424440.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000433428.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000440792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000452881.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000454918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000455816.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | ENST00000612823.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | NM_139073.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200179257.hg.1 | TC0200011048.hg.1 | Clariom D TC |

SPATA3
Genomic Map