| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025637.hg.1 |
ENST00000354300.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025637.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12025637.hg.1 |
TC12001966.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200192539.hg.1 |
TC1200011911.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000354300.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
ENST00000548721.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011911.hg.1 |
NM_139283.1 |
Refseq |