| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
ENST00000232125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
ENST00000440333.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
ENST00000477892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011371.hg.1 |
TC03000631.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
ENST00000232125.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
ENST00000440333.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
ENST00000477892.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03011370.hg.1 |
TC03000631.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300008551.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000232125.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000469967.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
ENST00000477892.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
NM_014367.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300160857.hg.1 |
TC0300008551.hg.1 |
Clariom D TC |