Assay Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK056041.1 | - | 3 - 4 | 570 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK056041.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
