Assay Details
| Gene Symbol: DRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145038.4 | NP_659475.2 | 6 - 7 | 846 | 74 | |
| XM_005264637.3 | XP_005264694.1 | 5 - 6 | 654 | 74 | ||
| XM_005264638.3 | XP_005264695.1 | 3 - 4 | 341 | 74 | ||
| XM_017005271.1 | XP_016860760.1 | 6 - 7 | 754 | 74 | ||
| GenBank mRNA | AK057222.1 | - | 6 - 7 | 842 | 74 | |
| AK289953.1 | - | 6 - 7 | 829 | 74 | ||
| AK307094.1 | - | 5 - 6 | 636 | 74 | ||
| AL833892.1 | - | 5 - 6 | 580 | 74 | ||
| BC009488.2 | - | 5 - 6 | 569 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000151.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005264638.3,XM_017005271.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001786.hg.1 | ENST00000288710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001786.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001786.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001786.hg.1 | TC02000151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147887.hg.1 | TC0200007038.hg.1 | Clariom D TC |

DRC1
Genomic Map