Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304458.1 | NP_001291387.1 | 5 - 6 | 1043 | 75 | |
| NM_138364.3 | NP_612373.2 | 6 - 7 | 1192 | 75 | ||
| XM_017008823.1 | XP_016864312.1 | 6 - 7 | 933 | 75 | ||
| XM_017008824.1 | XP_016864313.1 | 5 - 6 | 865 | 75 | ||
| XM_017008825.1 | XP_016864314.1 | 5 - 6 | 919 | 75 | ||
| XM_017008826.1 | XP_016864315.1 | 4 - 5 | 814 | 75 | ||
| GenBank mRNA | AK096703.1 | - | 5 - 6 | 938 | 75 | |
| AK292904.1 | - | 6 - 7 | 1125 | 75 | ||
| BC021250.2 | - | 6 - 7 | 947 | 75 | ||
| BC064403.1 | - | 6 - 7 | 1091 | 5547611 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001625.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017008823.1,XM_017008824.1,XM_017008825.1,XM_017008826.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04022686.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022686.hg.1 | ENST00000541232.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022686.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022686.hg.1 | TC04001625.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000511687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012068.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000322396.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000510269.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | ENST00000514886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | NM_001304458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | NM_138364.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182883.hg.1 | TC0400012068.hg.1 | Clariom D TC |

PRMT9
Genomic Map