| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000461075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000461075.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000474677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000474677.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020849.hg.1 |
TC0X001556.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020849.hg.1 |
ENST00000334398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020849.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020849.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X020849.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00176949.hg.1 |
TC0X00011248.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000334398.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000369439.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000449645.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000461075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000461075.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000474677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000474677.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
ENST00000487422.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_001184797.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011248.hg.1 |
NM_018196.3 |
Refseq |