| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011819.hg.1 |
ENST00000301744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011819.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011819.hg.1 |
TC16000817.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011818.hg.1 |
ENST00000301744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011818.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011818.hg.1 |
TC16000817.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011820.hg.1 |
ENST00000301744.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011820.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16011820.hg.1 |
TC16000817.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009194.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170172.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170172.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170172.hg.1 |
TC1600009194.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170172.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170172.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170175.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170175.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170175.hg.1 |
TC1600009194.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170175.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170175.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170173.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170173.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170173.hg.1 |
TC1600009194.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170173.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170173.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170174.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170174.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170174.hg.1 |
TC1600009194.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170174.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170174.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170171.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170171.hg.1 |
NM_152457.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600170171.hg.1 |
TC1600009194.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600170171.hg.1 |
ENST00000301744.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600170171.hg.1 |
NM_152457.2 |
Refseq |